missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorophenol 98.0+%, TCI America™

Catalog No. p-7127066
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20155G 5 g
D201525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20155G Supplier TCI America Supplier No. D20155G
Only null left

Chemical Identifiers

CAS 28177-48-2
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.094
MDL Number MFCD00002158
InChI Key CKKOVFGIBXCEIJ-UHFFFAOYSA-N
Synonym phenol, 2,6-difluoro, 2.6-difluorophenol, 2,6-difluoro-phenol, 2,6-difluorophenol, 2,6-difluoro phenol, pubchem1495, ksc493c1n, ckkovfgibxceij-uhfffaoysa
PubChem CID 94392
IUPAC Name 2,6-difluorophenol
SMILES C1=CC(=C(C(=C1)F)O)F

Specifications

Melting Point 45°C
Color White
Boiling Point 60°C
Quantity 5 g
UN Number 1325
Formula Weight 130.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Difluorophenol
TSCA No
Recommended Storage Refrigerator