missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoronitrobenzene 98.0+%, TCI America™

Catalog No. d41235g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41235G 5 g
D41231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41235G Supplier TCI America Supplier No. D41235G
Only null left

Chemical Identifiers

CAS 19064-24-5
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.092
MDL Number MFCD00192035
InChI Key SSNCMIDZGFCTST-UHFFFAOYSA-N
Synonym 2,6-difluoronitrobenzene, 1,3-difluoro-2-nitro-benzene, benzene, 1,3-difluoro-2-nitro, 2,6-difluoro nitrobenzene, pubchem4403, 1,3-difluoronitrobenzene, acmc-1buv1, 2,6-difluoro-nitrobenzene, 2,6-difluoro nitro benzene, 1,3-difluoro-2-nitrobenzen
PubChem CID 87922
IUPAC Name 1,3-difluoro-2-nitrobenzene
SMILES C1=CC(=C(C(=C1)F)[N+](=O)[O-])F

Specifications

Boiling Point 92°C
Quantity 5 g
Formula Weight 159.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,6-Difluoronitrobenzene