accessibility menu, dialog, popup

UserName

CAS RN 19064-24-5

IUPAC Name: 1,3-difluoro-2-nitrobenzene
Synonyms: 1,3-difluoro-2-nitrobenzene
Molecular Weight (g/mol): 159.09
Molecular Formula: C6H3F2NO2
InChi Key: SSNCMIDZGFCTST-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(F)C=CC=C1F
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)