missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluorobenzylamine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 177°C |
| Quantity | 5 g |
| UN Number | 2735 |
| Synonym | 2,6-difluorobenzylamine, 2,6-difluorophenyl methanamine, 2,6-difluorobenzyl amine, benzenemethanamine, 2,6-difluoro, 1-2,6-difluorophenyl methanamine, 2,6-difluorophenyl methylamine, 2, 6-difluorobenzylamine, pubchem2235, 2,6-difluoro-benzylamine, acmc-1b2iy |
| Formula Weight | 143.14 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,6-Difluorobenzylamine |
Safety and Handling
| TSCA | No |