CAS RN 69385-30-4
IUPAC Name:
(2,6-difluorophenyl)methanaminium
Synonyms:
(2,6-difluorophenyl)methanaminium
Molecular Weight (g/mol):
144.14
Molecular Formula:
C7H8F2N
InChi Key:
PQCUDKMMPTXMAL-UHFFFAOYSA-O
SMILES:
[NH3+]CC1=C(F)C=CC=C1F
1
–
1
of
1
results
1
2,6-Difluorobenzylamine 98.0+%, TCI America™
CAS: 69385-30-4 Molecular Formula: C7H8F2N Molecular Weight (g/mol): 144.14 MDL Number: MFCD00010144 InChI Key: PQCUDKMMPTXMAL-UHFFFAOYSA-O PubChem CID: 123563 IUPAC Name: (2,6-difluorophenyl)methanaminium SMILES: [NH3+]CC1=C(F)C=CC=C1F