missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzyl Cyanide 98.0+%, TCI America™

Catalog No. D45105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45105G 5 g
D451025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45105G Supplier TCI America Supplier No. D45105G
Only null left

Chemical Identifiers

CAS 654-01-3
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.13
MDL Number MFCD00009973
InChI Key GVAYBGQTAADLJS-UHFFFAOYSA-N
Synonym 2,6-difluorophenylacetonitrile, 2-2,6-difluorophenyl acetonitrile, 2,6-difluorobenzyl cyanide, 2,6-difluorophenyl acetonitrile, 2,6-difluorobenzeneacetonitrile, benzeneacetonitrile, 2,6-difluoro, 2-2,6-difluorophenyl ethanenitrile, pubchem4152, acmc-1axnw, n-dodecyl-beta-maltoside
PubChem CID 69559
IUPAC Name 2-(2,6-difluorophenyl)acetonitrile
SMILES FC1=CC=CC(F)=C1CC#N

Specifications

Color Yellow
Boiling Point 109°C
Quantity 5 g
UN Number 1992
Formula Weight 153.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Difluorobenzyl Cyanide