CAS RN 654-01-3
IUPAC Name:
2-(2,6-difluorophenyl)acetonitrile
Synonyms:
2-(2,6-difluorophenyl)acetonitrile
Molecular Weight (g/mol):
153.13
Molecular Formula:
C8H5F2N
InChi Key:
GVAYBGQTAADLJS-UHFFFAOYSA-N
SMILES:
FC1=CC=CC(F)=C1CC#N
1
–
1
of
1
results
1
2,6-Difluorobenzyl Cyanide 98.0+%, TCI America™
CAS: 654-01-3 Molecular Formula: C8H5F2N Molecular Weight (g/mol): 153.13 MDL Number: MFCD00009973 InChI Key: GVAYBGQTAADLJS-UHFFFAOYSA-N Synonym: 2,6-difluorophenylacetonitrile,2-2,6-difluorophenyl acetonitrile,2,6-difluorobenzyl cyanide,2,6-difluorophenyl acetonitrile,2,6-difluorobenzeneacetonitrile,benzeneacetonitrile, 2,6-difluoro,2-2,6-difluorophenyl ethanenitrile,pubchem4152,acmc-1axnw,n-dodecyl-beta-maltoside PubChem CID: 69559 IUPAC Name: 2-(2,6-difluorophenyl)acetonitrile SMILES: FC1=CC=CC(F)=C1CC#N