missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzoyl chloride, 98%

Catalog No. AAA1128606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1128606 5 g
AAA1128614 25 g
AAA1128622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1128606 Supplier Thermo Scientific Chemicals Supplier No. A1128606
Only null left

CAS: 18063-02-0 | C7H3ClF2O | 176.547 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18063-02-0
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD00000659
InChI Key QRHUZEVERIHEPT-UHFFFAOYSA-N
Synonym 2,6-difluorobenzoylchloride, benzoyl chloride, 2,6-difluoro, 2,6-difluoro-benzoyl chloride, acmc-1ca8w, 2,6-diflurobenzoyl chloride, 2 6-difluorobenzoyl chloride, 2,6-diflourobenzoyl chloride, 2,6-difluoro benzoylchloride, 2.6-difluorobenzoyl chloride, 2,6-difluro-benzoyl chloride
PubChem CID 87438
IUPAC Name 2,6-difluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1)F)C(=O)Cl)F

Specifications

Density 1.407
Boiling Point 72°C to 77°C (13 mmHg)
Flash Point 72°C (162°F)
Refractive Index 1.4988
Quantity 5 g
UN Number UN3265
Beilstein 639438
Sensitivity Moisture sensitive
Formula Weight 176.55
Percent Purity 98%
Chemical Name or Material 2,6-Difluorobenzoyl chloride
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 241-971-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only