accessibility menu, dialog, popup

UserName

FFClO

CAS RN 18063-02-0

IUPAC Name: 2,6-difluorobenzoyl chloride
Synonyms: 2,6-difluorobenzoyl chloride
Molecular Weight (g/mol): 176.55
Molecular Formula: C7H3ClF2O
InChi Key: QRHUZEVERIHEPT-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1C(Cl)=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)