missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluorobenzonitrile 99.0+%, TCI America™
$42.87 - $115.57
Chemical Identifiers
| CAS | 1897-52-5 |
|---|---|
| Molecular Formula | C7H3F2N |
| Molecular Weight (g/mol) | 139.105 |
| MDL Number | MFCD00001778 |
| InChI Key | BNBRIFIJRKJGEI-UHFFFAOYSA-N |
| Synonym | 2,6-difluorobenznitrile, benzonitrile, 2,6-difluoro, 2,6-difluoro-benzonitrile, 2,6-dfbn, unii-1j58z42a2e, 2,6-difluoro benzonitrile, 2,6-difluorobenzenecarbonitrile, 2,6-difluorophenylcyanide, ncr bf ff, 6-difluorobenzonitrile |
| PubChem CID | 74695 |
| IUPAC Name | 2,6-difluorobenzonitrile |
| SMILES | C1=CC(=C(C(=C1)F)C#N)F |
Chemical Identifiers
| 1897-52-5 | |
| 139.105 | |
| BNBRIFIJRKJGEI-UHFFFAOYSA-N | |
| 74695 | |
| C1=CC(=C(C(=C1)F)C#N)F |
| C7H3F2N | |
| MFCD00001778 | |
| 2,6-difluorobenznitrile, benzonitrile, 2,6-difluoro, 2,6-difluoro-benzonitrile, 2,6-dfbn, unii-1j58z42a2e, 2,6-difluoro benzonitrile, 2,6-difluorobenzenecarbonitrile, 2,6-difluorophenylcyanide, ncr bf ff, 6-difluorobenzonitrile | |
| 2,6-difluorobenzonitrile |
Specifications
| 1897-52-5 | |
| C7H3F2N | |
| 25 g | |
| 2,6-difluorobenznitrile, benzonitrile, 2,6-difluoro, 2,6-difluoro-benzonitrile, 2,6-dfbn, unii-1j58z42a2e, 2,6-difluoro benzonitrile, 2,6-difluorobenzenecarbonitrile, 2,6-difluorophenylcyanide, ncr bf ff, 6-difluorobenzonitrile | |
| C1=CC(=C(C(=C1)F)C#N)F | |
| 139.105 | |
| 139.10 | |
| Crystalline Lumps |
| White | |
| MFCD00001778 | |
| 3439 | |
| BNBRIFIJRKJGEI-UHFFFAOYSA-N | |
| 2,6-difluorobenzonitrile | |
| 74695 | |
| ≥99.0% (GC) | |
| 2,6-Difluorobenzonitrile |
Safety and Handling
EINECSNumber : (3)-3854
TSCA : Yes