accessibility menu, dialog, popup

UserName

FFN

CAS RN 1897-52-5

IUPAC Name: 2,6-difluorobenzonitrile
Synonyms: 2,6-difluorobenzonitrile
Molecular Weight (g/mol): 139.11
Molecular Formula: C7H3F2N
InChi Key: BNBRIFIJRKJGEI-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)