missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzonitrile 99.0+%, TCI America™

Catalog No. D165025G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D165025G 25 g
D1650100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D165025G Supplier TCI America Supplier No. D165025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1897-52-5
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.105
MDL Number MFCD00001778
InChI Key BNBRIFIJRKJGEI-UHFFFAOYSA-N
Synonym 2,6-difluorobenznitrile, benzonitrile, 2,6-difluoro, 2,6-difluoro-benzonitrile, 2,6-dfbn, unii-1j58z42a2e, 2,6-difluoro benzonitrile, 2,6-difluorobenzenecarbonitrile, 2,6-difluorophenylcyanide, ncr bf ff, 6-difluorobenzonitrile
PubChem CID 74695
IUPAC Name 2,6-difluorobenzonitrile
SMILES C1=CC(=C(C(=C1)F)C#N)F

Specifications

Color White
Quantity 25 g
UN Number 3439
Formula Weight 139.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,6-Difluorobenzonitrile
EINECSNumber (3)-3854
TSCA Yes