missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoroanisole 97.0+%, TCI America™

Catalog No. D33605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33605G 5 g
D33601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33605G Supplier TCI America Supplier No. D33605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 437-82-1
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
InChI Key IOBWAHRFIPQEQL-UHFFFAOYSA-N
Synonym 2,6-difluoroanisole, 1,3-difluoro-2-methoxy-benzene, benzene, 1,3-difluoro-2-methoxy, pubchem1968, 2,6-difluoro anisole, 2.6-difluoro anisole, acmc-1ajjj, ksc235o3h, iobwahrfipqeql-uhfffaoysa
PubChem CID 2736897
IUPAC Name 1,3-difluoro-2-methoxybenzene
SMILES COC1=C(C=CC=C1F)F

Specifications

Color Yellow
Boiling Point 50°C
Quantity 5 g
UN Number 1993
Formula Weight 144.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Difluoroanisole