accessibility menu, dialog, popup

UserName

CAS RN 437-82-1

H3COFF

CAS RN 437-82-1

IUPAC Name: 1,3-difluoro-2-methoxybenzene
Synonyms: 1,3-difluoro-2-methoxybenzene
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: IOBWAHRFIPQEQL-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results