missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluoroaniline 98.0+%, TCI America™
Supplier: TCI America D1635100G
Specifications
| 2,6-Difluoroaniline | |
| 154°C | |
| MFCD00007655 | |
| 1992 | |
| ODUZJBKKYBQIBX-UHFFFAOYSA-N | |
| 2,6-difluoroaniline | |
| 79647 | |
| ≥98.0% (GC) |
| 5509-65-9 | |
| C6H5F2N | |
| 100 g | |
| benzenamine, 2,6-difluoro, 2,6-difluoro-phenylamine, 2,6-difluoro aniline, 2,6-difluorobenzenamine, 2,6-difluoranilin, 2,6 difluoroaniline, 2,6-diflouroaniline, pubchem4034, 2,6-difuluoroaniline | |
| NC1=C(F)C=CC=C1F | |
| 129.11 | |
| 129.11 | |
| Liquid |
Chemical Identifiers
| 5509-65-9 | |
| 129.11 | |
| ODUZJBKKYBQIBX-UHFFFAOYSA-N | |
| 79647 | |
| NC1=C(F)C=CC=C1F |
| C6H5F2N | |
| MFCD00007655 | |
| benzenamine, 2,6-difluoro, 2,6-difluoro-phenylamine, 2,6-difluoro aniline, 2,6-difluorobenzenamine, 2,6-difluoranilin, 2,6 difluoroaniline, 2,6-diflouroaniline, pubchem4034, 2,6-difuluoroaniline | |
| 2,6-difluoroaniline |
Safety and Handling
EINECSNumber : (3)-4104
TSCA : No