missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoroaniline 98.0+%, TCI America™

Catalog No. p-7126840
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16355G 5 g
D163525G 25 g
D1635100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D16355G Supplier TCI America Supplier No. D16355G
Only null left

Chemical Identifiers

CAS 5509-65-9
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007655
InChI Key ODUZJBKKYBQIBX-UHFFFAOYSA-N
Synonym benzenamine, 2,6-difluoro, 2,6-difluoro-phenylamine, 2,6-difluoro aniline, 2,6-difluorobenzenamine, 2,6-difluoranilin, 2,6 difluoroaniline, 2,6-diflouroaniline, pubchem4034, 2,6-difuluoroaniline
PubChem CID 79647
IUPAC Name 2,6-difluoroaniline
SMILES NC1=C(F)C=CC=C1F

Specifications

Boiling Point 154°C
Quantity 5 g
UN Number 1992
Formula Weight 129.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Difluoroaniline
EINECSNumber (3)-4104
TSCA No