missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylhydrazine hydrochloride, 98+%

Catalog No. AAA1196803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1196803 1 g
AAA1196806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1196803 Supplier Thermo Scientific Chemicals Supplier No. A1196803

CAS: 50709-36-9 | C6H7Cl3N2 | 213.486 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50709-36-9
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
MDL Number MFCD00012930
InChI Key CQNIYLLTIOPFCJ-UHFFFAOYSA-N
Synonym 2,6-dichlorophenylhydrazine hydrochloride, 2,6-dichlorophenyl hydrazine hydrochloride, 1-2,6-dichlorophenyl hydrazine hydrochloride, 2,6-dichlorophenyl hydrazine monohydrochloride, hydrazine, 2,6-dichlorophenyl-, monohydrochloride, 2,6-dichlorophenylhydrazinehydrochloride, acmc-209kpe, ksc491e8j, 2,6-dichlorophenylhydrazine, chloride, 2,6-dichlorophenylhydrazine hcl
PubChem CID 2723909
IUPAC Name (2,6-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=C(C(=C1)Cl)NN)Cl.Cl

Specifications

Melting Point ∼225°C (decomposition)
Quantity 1 g
Beilstein 4569738
Formula Weight 213.5
Percent Purity ≥98%
Chemical Name or Material 2,6-Dichlorophenylhydrazine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H301-H311-H332-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 256-730-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only