missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzyl Methyl Ether, TCI America™

Catalog No. D210925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D210925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D210925G Supplier TCI America Supplier No. D210925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33486-90-7
Molecular Formula C8H8Cl2O
Molecular Weight (g/mol) 191.05
MDL Number MFCD00000578
InChI Key QBKBHXIQLAMKOB-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzyl methyl ether, 1,3-dichloro-2-methoxymethyl benzene, benzene, 1,3-dichloro-2-methoxymethyl, 2,6-dichlorobenzylmethylether, acmc-1afwi, qbkbhxiqlamkob-uhfffaoysa, 2,6-dichloro-alpha-methoxytoluene, 2,6-dichlorophenyl methoxymethane, 1,3-dichloro-2-methoxymethyl-benzene, 1,3-dichloro-2-methoxymethyl benzene #
PubChem CID 118490
IUPAC Name 1,3-dichloro-2-(methoxymethyl)benzene
SMILES COCC1=C(Cl)C=CC=C1Cl

Specifications

Color Colorless
Boiling Point 125°C
Quantity 25 g
Formula Weight 191.05
Physical Form Liquid
Chemical Name or Material 2,6-Dichlorobenzyl Methyl Ether