CAS RN 33486-90-7
IUPAC Name:
1,3-dichloro-2-(methoxymethyl)benzene
Synonyms:
1,3-dichloro-2-(methoxymethyl)benzene
Molecular Weight (g/mol):
191.05
Molecular Formula:
C8H8Cl2O
InChi Key:
QBKBHXIQLAMKOB-UHFFFAOYSA-N
SMILES:
COCC1=C(Cl)C=CC=C1Cl
1
–
1
of
1
results
1
2,6-Dichlorobenzyl Methyl Ether, TCI America™
CAS: 33486-90-7 Molecular Formula: C8H8Cl2O Molecular Weight (g/mol): 191.05 MDL Number: MFCD00000578 InChI Key: QBKBHXIQLAMKOB-UHFFFAOYSA-N Synonym: 2,6-dichlorobenzyl methyl ether,1,3-dichloro-2-methoxymethyl benzene,benzene, 1,3-dichloro-2-methoxymethyl,2,6-dichlorobenzylmethylether,acmc-1afwi,qbkbhxiqlamkob-uhfffaoysa,2,6-dichloro-alpha-methoxytoluene,2,6-dichlorophenyl methoxymethane,1,3-dichloro-2-methoxymethyl-benzene,1,3-dichloro-2-methoxymethyl benzene # PubChem CID: 118490 IUPAC Name: 1,3-dichloro-2-(methoxymethyl)benzene SMILES: COCC1=C(Cl)C=CC=C1Cl