missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorobenzoic Acid 98.0+%, TCI America™
Supplier: TCI America D03395G
Specifications
| 2,6-Dichlorobenzoic Acid | |
| 144°C | |
| C7H4Cl2O2 | |
| 5 g | |
| MRUDNSFOFOQZDA-UHFFFAOYSA-N | |
| 2,6-dichlorobenzoic acid | |
| 5758 | |
| 191.01 | |
| Crystalline Powder |
| 50-30-6 | |
| White | |
| MFCD00002418 | |
| benzoic acid, 2,6-dichloro, 2,6-dichloro-benzoic acid, unii-634ri764qt, 2,6 dichlorobenzoic acid, 2,6-dichlorobenzoicacid, 2,6-dichloro-benzoicacid, ccris 8610, pubchem20028, acmc-1auop, 2.6-dichlorobenzoic acid | |
| OC(=O)C1=C(Cl)C=CC=C1Cl | |
| 191.01 | |
| CHEBI:48623 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 50-30-6 | |
| 191.01 | |
| MRUDNSFOFOQZDA-UHFFFAOYSA-N | |
| 5758 | |
| 2,6-dichlorobenzoic acid |
| C7H4Cl2O2 | |
| MFCD00002418 | |
| benzoic acid, 2,6-dichloro, 2,6-dichloro-benzoic acid, unii-634ri764qt, 2,6 dichlorobenzoic acid, 2,6-dichlorobenzoicacid, 2,6-dichloro-benzoicacid, ccris 8610, pubchem20028, acmc-1auop, 2.6-dichlorobenzoic acid | |
| CHEBI:48623 | |
| OC(=O)C1=C(Cl)C=CC=C1Cl |
Safety and Handling
EINECSNumber : (3)-1410&(3)-1424
RTECSNumber : DG7000000
TSCA : No