missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7126156
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D03395G 5 g
D033925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D03395G Supplier TCI America Supplier No. D03395G
Only null left

Chemical Identifiers

CAS 50-30-6
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002418
InChI Key MRUDNSFOFOQZDA-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-dichloro, 2,6-dichloro-benzoic acid, unii-634ri764qt, 2,6 dichlorobenzoic acid, 2,6-dichlorobenzoicacid, 2,6-dichloro-benzoicacid, ccris 8610, pubchem20028, acmc-1auop, 2.6-dichlorobenzoic acid
PubChem CID 5758
ChEBI CHEBI:48623
IUPAC Name 2,6-dichlorobenzoic acid
SMILES OC(=O)C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 144°C
Color White
Quantity 5 g
Formula Weight 191.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorobenzoic Acid
EINECSNumber (3)-1410&(3)-1424
RTECSNumber DG7000000
TSCA No