missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloro-4-fluoroaniline 97.0+%, TCI America™
$59.65 - $175.22
Chemical Identifiers
| CAS | 344-19-4 |
|---|---|
| Molecular Formula | C6H4Cl2FN |
| Molecular Weight (g/mol) | 180.003 |
| MDL Number | MFCD00142845 |
| InChI Key | YAUYKCFMKMZTEX-UHFFFAOYSA-N |
| Synonym | 2,6-dichloro-4-fluorobenzenamine, 2,6-dichloro-4-fluoro-aniline, 2',6'-dichloro-4'-fluoroaniline, benzenamine, 2,6-dichloro-4-fluoro, 2,6-dichloro-4-fluorophenylamine, pubchem3451, acmc-1ad5c, ksc572g5t, 2,6,-dichloro-4-fluoroaniline, 2,6 dichloro-4-fluorophenylamine |
| PubChem CID | 2774008 |
| IUPAC Name | 2,6-dichloro-4-fluoroaniline |
| SMILES | C1=C(C=C(C(=C1Cl)N)Cl)F |
Chemical Identifiers
| 344-19-4 | |
| 180.003 | |
| YAUYKCFMKMZTEX-UHFFFAOYSA-N | |
| 2774008 | |
| C1=C(C=C(C(=C1Cl)N)Cl)F |
| C6H4Cl2FN | |
| MFCD00142845 | |
| 2,6-dichloro-4-fluorobenzenamine, 2,6-dichloro-4-fluoro-aniline, 2',6'-dichloro-4'-fluoroaniline, benzenamine, 2,6-dichloro-4-fluoro, 2,6-dichloro-4-fluorophenylamine, pubchem3451, acmc-1ad5c, ksc572g5t, 2,6,-dichloro-4-fluoroaniline, 2,6 dichloro-4-fluorophenylamine | |
| 2,6-dichloro-4-fluoroaniline |
Specifications
| 344-19-4 | |
| C6H4Cl2FN | |
| 1 g | |
| 2,6-dichloro-4-fluorobenzenamine, 2,6-dichloro-4-fluoro-aniline, 2',6'-dichloro-4'-fluoroaniline, benzenamine, 2,6-dichloro-4-fluoro, 2,6-dichloro-4-fluorophenylamine, pubchem3451, acmc-1ad5c, ksc572g5t, 2,6,-dichloro-4-fluoroaniline, 2,6 dichloro-4-fluorophenylamine | |
| C1=C(C=C(C(=C1Cl)N)Cl)F | |
| 180.003 | |
| 180.00 | |
| Crystalline Powder |
| 56°C | |
| MFCD00142845 | |
| 2811 | |
| YAUYKCFMKMZTEX-UHFFFAOYSA-N | |
| 2,6-dichloro-4-fluoroaniline | |
| 2774008 | |
| ≥97.0% (GC,N) | |
| 2,6-Dichloro-4-fluoroaniline |
Safety and Handling
TSCA : No