accessibility menu, dialog, popup

UserName

CAS RN 344-19-4

ClH2NClF

CAS RN 344-19-4

IUPAC Name: 2,6-dichloro-4-fluoroaniline
Synonyms: 2,6-dichloro-4-fluoroaniline
Molecular Weight (g/mol): 180
Molecular Formula: C6H4Cl2FN
InChi Key: YAUYKCFMKMZTEX-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(F)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results