missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97.0+%, TCI America™

Catalog No. p-7128151
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42011G 1 g
D42015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42011G Supplier TCI America Supplier No. D42011G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1003298-87-0
Molecular Formula C12H15BCl2O3
Molecular Weight (g/mol) 288.959
MDL Number MFCD20526385
InChI Key IAVSUBSFIDLDNW-UHFFFAOYSA-N
Synonym 3,5-Dichloro-4-hydroxyphenylboronic Acid Pinacol Ester, 2-(3,5-Dichloro-4-hydroxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 66715127
IUPAC Name 2,6-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)Cl)O)Cl

Specifications

Melting Point 110°C
Color White
Boiling Point 103°C
Quantity 1 g
Formula Weight 288.96
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol