accessibility menu, dialog, popup

UserName

CAS RN 1003298-87-0

IUPAC Name: 2,6-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Synonyms: 2,6-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular Weight (g/mol): 288.96
Molecular Formula: C12H15BCl2O3
InChi Key: IAVSUBSFIDLDNW-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(Cl)=C(O)C(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results