missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromophenol 98.0+%, TCI America™

Catalog No. p-7127706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33845G 5 g
D338425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33845G Supplier TCI America Supplier No. D33845G
Only null left

Chemical Identifiers

CAS 608-33-3
Molecular Formula C6H4Br2O
Molecular Weight (g/mol) 251.91
MDL Number MFCD00002152
InChI Key SSIZLKDLDKIHEV-UHFFFAOYSA-N
Synonym phenol, 2,6-dibromo, 2,6-dibromo-phenol, unii-27mip05eav, 2,6-dibromo-pheno, 27mip05eav, 2,6-bis bromanyl phenol, acmc-1au9i, 2,6-dibromophenol, ksc493e5t, 2,6-dibromophenol, analytical standard
PubChem CID 11847
ChEBI CHEBI:19391
IUPAC Name 2,6-dibromophenol
SMILES OC1=C(Br)C=CC=C1Br

Specifications

Melting Point 56°C
Boiling Point 256°C
Quantity 5 g
UN Number 2811
Formula Weight 251.91
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromophenol