accessibility menu, dialog, popup

UserName

OHBrBr

CAS RN 608-33-3

IUPAC Name: 2,6-dibromophenol
Synonyms: 2,6-dibromophenol
Molecular Weight (g/mol): 251.91
Molecular Formula: C6H4Br2O
InChi Key: SSIZLKDLDKIHEV-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=CC=C1Br
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)