missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromoanthracene 98.0+%, TCI America™
$654.29 - $2,223.83
Chemical Identifiers
| CAS | 186517-01-1 |
|---|---|
| Molecular Formula | C14H8Br2 |
| Molecular Weight (g/mol) | 336.026 |
| InChI Key | BPRGLVVFWRNXEP-UHFFFAOYSA-N |
| PubChem CID | 22058906 |
| IUPAC Name | 2,6-dibromoanthracene |
| SMILES | C1=CC(=CC2=CC3=C(C=C21)C=C(C=C3)Br)Br |
Chemical Identifiers
| 186517-01-1 | |
| 336.026 | |
| 22058906 | |
| C1=CC(=CC2=CC3=C(C=C21)C=C(C=C3)Br)Br |
| C14H8Br2 | |
| BPRGLVVFWRNXEP-UHFFFAOYSA-N | |
| 2,6-dibromoanthracene |
Specifications
| 186517-01-1 | |
| 1 g | |
| C1=CC(=CC2=CC3=C(C=C21)C=C(C=C3)Br)Br | |
| 336.026 | |
| 336.03 | |
| Crystalline Powder |
| C14H8Br2 | |
| BPRGLVVFWRNXEP-UHFFFAOYSA-N | |
| 2,6-dibromoanthracene | |
| 22058906 | |
| ≥98.0% (HPLC) | |
| 2,6-Dibromoanthracene |
Safety and Handling
TSCA : No