missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-methylaniline 98.0+%, TCI America™

Catalog No. p-7127799
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35775G 5 g
D357725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35775G Supplier TCI America Supplier No. D35775G
Only null left

Chemical Identifiers

CAS 6968-24-7
Molecular Formula C7H7Br2N
Molecular Weight (g/mol) 264.948
MDL Number MFCD00007641
InChI Key ATDIROHVRVQMRO-UHFFFAOYSA-N
Synonym 2,6-dibromo-p-toluidine, 3,5-dibromo-4-aminotoluene, benzenamine, 2,6-dibromo-4-methyl, 2,6-dibromo-4-methyl aniline, 2,6-dibromo-4-methyl-phenylamine, 2,6-dibromo-4-methylphenylamine, 2,6-dibromo-4-methyl-benzenamine, pubchem2334, acmc-209oag, 2,6-dibromo-p-toludine
PubChem CID 81427
IUPAC Name 2,6-dibromo-4-methylaniline
SMILES CC1=CC(=C(C(=C1)Br)N)Br

Specifications

Melting Point 74°C
Quantity 5 g
Formula Weight 264.95
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromo-4-methylaniline