missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Diaminopimelic Acid 98.0+%, TCI America™
Supplier: TCI America D01125G
Specifications
| 2,6-Diaminopimelic Acid | |
| White-Yellow | |
| MFCD00002637 | |
| 2,6-diaminopimelic acid, heptanedioic acid, 2,6-diamino, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, m-dap, dl-alpha,epsilon-diaminopimelic acid, 1,5-diaminoheptanedioic acid, ll-diaminopimelate, d,l-diaminopimelate, ll-a2pm | |
| C(CC(C(=O)O)N)CC(C(=O)O)N | |
| 190.199 | |
| CHEBI:23673 | |
| ≥98.0% (T) |
| 583-93-7 | |
| C7H14N2O4 | |
| 5 g | |
| GMKMEZVLHJARHF-UHFFFAOYSA-N | |
| 2,6-diaminoheptanedioic acid | |
| 865 | |
| 190.20 | |
| Crystalline Powder |
Chemical Identifiers
| 583-93-7 | |
| 190.199 | |
| GMKMEZVLHJARHF-UHFFFAOYSA-N | |
| 865 | |
| 2,6-diaminoheptanedioic acid |
| C7H14N2O4 | |
| MFCD00002637 | |
| 2,6-diaminopimelic acid, heptanedioic acid, 2,6-diamino, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, m-dap, dl-alpha,epsilon-diaminopimelic acid, 1,5-diaminoheptanedioic acid, ll-diaminopimelate, d,l-diaminopimelate, ll-a2pm | |
| CHEBI:23673 | |
| C(CC(C(=O)O)N)CC(C(=O)O)N |
Safety and Handling
EINECSNumber : (9)-0548
TSCA : No