accessibility menu, dialog, popup

UserName

NH3(S)(R)NH3OOOO

CAS RN 583-93-7

IUPAC Name: (2R,6S)-2,6-diazaniumylheptanedioate
Synonyms: meso-2,6-diaminopimelate
Molecular Weight (g/mol): 190.2
Molecular Formula: C7H14N2O4
InChi Key: GMKMEZVLHJARHF-SYDPRGILSA-N
SMILES: [NH3+][C@@H](CCC[C@@H]([NH3+])C([O-])=O)C([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)