missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminopimelic Acid 98.0+%, TCI America™

Catalog No. p-7126041
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01121G 1 g
D01125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D01121G Supplier TCI America Supplier No. D01121G
Only null left

Chemical Identifiers

CAS 583-93-7
Molecular Formula C7H14N2O4
Molecular Weight (g/mol) 190.199
MDL Number MFCD00002637
InChI Key GMKMEZVLHJARHF-UHFFFAOYSA-N
Synonym 2,6-diaminopimelic acid, heptanedioic acid, 2,6-diamino, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, m-dap, dl-alpha,epsilon-diaminopimelic acid, 1,5-diaminoheptanedioic acid, ll-diaminopimelate, d,l-diaminopimelate, ll-a2pm
PubChem CID 865
ChEBI CHEBI:23673
IUPAC Name 2,6-diaminoheptanedioic acid
SMILES C(CC(C(=O)O)N)CC(C(=O)O)N

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 190.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diaminopimelic Acid
EINECSNumber (9)-0548
TSCA No