missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-p-cresol 99.0+%, TCI America™

Supplier:  TCI America D0228500G

 View more versions of this product

Catalog No. D0228500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,6-Di-tert-butyl-p-cresol
71°C
265°C
MFCD00011644
3077
NLZUEZXRPGMBCV-UHFFFAOYSA-N
2,6-ditert-butyl-4-methylphenol
31404
220.36
Crystalline Powder
128-37-0
White
C15H24O
500 g
2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C
220.356
CHEBI:34247
≥99.0% (GC)

Chemical Identifiers

128-37-0
220.356
NLZUEZXRPGMBCV-UHFFFAOYSA-N
31404
2,6-ditert-butyl-4-methylphenol
C15H24O
MFCD00011644
2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
CHEBI:34247
CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0540&(9)-1805

RTECSNumber : GO7875000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions