missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-p-cresol 99.0+%, TCI America™

Catalog No. p-7126097
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D022825G 25 g
D0228500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D022825G Supplier TCI America Supplier No. D022825G
Only null left

Chemical Identifiers

CAS 128-37-0
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.356
MDL Number MFCD00011644
InChI Key NLZUEZXRPGMBCV-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
PubChem CID 31404
ChEBI CHEBI:34247
IUPAC Name 2,6-ditert-butyl-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

Melting Point 71°C
Color White
Boiling Point 265°C
Quantity 25 g
UN Number 3077
Formula Weight 220.36
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Di-tert-butyl-p-cresol
EINECSNumber (3)-0540&(9)-1805
RTECSNumber GO7875000
TSCA Yes