missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Di-tert-butyl-4-ethylphenol 98.0+%, TCI America™
$93.21 - $561.08
Chemical Identifiers
| CAS | 4130-42-1 |
|---|---|
| Molecular Formula | C16H26O |
| Molecular Weight (g/mol) | 234.38 |
| MDL Number | MFCD00017366 |
| InChI Key | BVUXDWXKPROUDO-UHFFFAOYSA-N |
| Synonym | 2,6-di-tert-butyl-4-ethylphenol, ionol 2, 4-ethyl-2,6-di-tert-butylphenol, phenol, 2,6-bis 1,1-dimethylethyl-4-ethyl, 1-hydroxy-4-ethyl-2,6-di-tert-butylbenzene, phenol, 2,6-di-tert-butyl-4-ethyl, unii-sv7oz6j97m, 2,6-bis 1,1-dimethylethyl-4-ethylphenol, yoshinox 250, sandant 425 |
| PubChem CID | 20087 |
| IUPAC Name | 2,6-di-tert-butyl-4-ethylphenol |
| SMILES | CCC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C |
Chemical Identifiers
| 4130-42-1 | |
| 234.38 | |
| BVUXDWXKPROUDO-UHFFFAOYSA-N | |
| 20087 | |
| CCC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C |
| C16H26O | |
| MFCD00017366 | |
| 2,6-di-tert-butyl-4-ethylphenol, ionol 2, 4-ethyl-2,6-di-tert-butylphenol, phenol, 2,6-bis 1,1-dimethylethyl-4-ethyl, 1-hydroxy-4-ethyl-2,6-di-tert-butylbenzene, phenol, 2,6-di-tert-butyl-4-ethyl, unii-sv7oz6j97m, 2,6-bis 1,1-dimethylethyl-4-ethylphenol, yoshinox 250, sandant 425 | |
| 2,6-di-tert-butyl-4-ethylphenol |
Specifications
| 4130-42-1 | |
| Yellow | |
| C16H26O | |
| 25 g | |
| 2,6-di-tert-butyl-4-ethylphenol, ionol 2, 4-ethyl-2,6-di-tert-butylphenol, phenol, 2,6-bis 1,1-dimethylethyl-4-ethyl, 1-hydroxy-4-ethyl-2,6-di-tert-butylbenzene, phenol, 2,6-di-tert-butyl-4-ethyl, unii-sv7oz6j97m, 2,6-bis 1,1-dimethylethyl-4-ethylphenol, yoshinox 250, sandant 425 | |
| CCC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C | |
| 234.38 | |
| 234.38 | |
| Crystalline Powder |
| 45°C | |
| 137°C | |
| MFCD00017366 | |
| 3077 | |
| BVUXDWXKPROUDO-UHFFFAOYSA-N | |
| 2,6-di-tert-butyl-4-ethylphenol | |
| 20087 | |
| ≥98.0% (GC) | |
| 2,6-Di-tert-butyl-4-ethylphenol |
Safety and Handling
EINECSNumber : (3)-0540
TSCA : Yes