missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Di-tert-butyl-4-ethylphenol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 45°C |
| Color | Yellow |
| Boiling Point | 137°C |
| Quantity | 25 g |
| UN Number | 3077 |
| Formula Weight | 234.38 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,6-Di-tert-butyl-4-ethylphenol |
Safety and Handling
| EINECSNumber | (3)-0540 |
| TSCA | Yes |