missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-dimethylaminomethylphenol 98.0+%, TCI America™

Catalog No. D230125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D230125G 25 g
D2301500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D230125G Supplier TCI America Supplier No. D230125G

Chemical Identifiers

CAS 88-27-7
Molecular Formula C17H29NO
Molecular Weight (g/mol) 263.425
MDL Number MFCD00026283
InChI Key VMZVBRIIHDRYGK-UHFFFAOYSA-N
Synonym ethyl 703, 2,6-di-tert-butyl-4-dimethylaminomethyl phenol, n,n-dimethyl-3,5-di-tert-butyl-4-hydroxybenzylamine, agidol 3, f 1 antioxidant, ethyl antioxidant 703, 2,6-di-tert-butyl-4-dimethylamino methylphenol, unii-8tk6vr2kyx, 2,6-di-tert-butyl-alpha-dimethylamino-p-cresol, n-3,5-di-tert-butyl-4-hydroxybenzyl dimethylamine
PubChem CID 66609
IUPAC Name 2,6-ditert-butyl-4-[(dimethylamino)methyl]phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CN(C)C

Specifications

Melting Point 94°C
Color White-Yellow
Boiling Point 179°C
Quantity 25 g
Formula Weight 263.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Di-tert-butyl-4-dimethylaminomethylphenol
EINECSNumber (3)-2865
RTECSNumber GO7887000
TSCA Yes