missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%

Catalog No. AAL19952MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19952MD 250 mg
AAL1995203 1 g
AAL1995206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL19952MD Supplier Thermo Scientific Chemicals Supplier No. L19952MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 681812-07-7 | C8H5BF6O2 | 257.926 g/mol

2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 681812-07-7
Molecular Formula C8H5BF6O2
Molecular Weight (g/mol) 257.926
MDL Number MFCD04039321
InChI Key WAMPGNNEOZGBHR-UHFFFAOYSA-N
Synonym 2,6-bis trifluoromethyl phenyl boronic acid, 2,6-bis trifluoromethyl phenylboronic acid, 2,6-bis trifluoromethyl benzeneboronic acid, 2,6-ditrifluoromethylphenyl boronic acid, boronic acid, 2,6-bis trifluoromethyl phenyl, acmc-1b92w, ablock ab-13-3574, 2,6-bis-trifluoromethyl benzeneboronic acid
PubChem CID 2783239
IUPAC Name [2,6-bis(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=CC=C1C(F)(F)F)C(F)(F)F)(O)O

Specifications

Melting Point ∼150°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 257.93
Percent Purity 97%
Chemical Name or Material 2,6-Bis(trifluoromethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only