Learn More
2,6-Bis(hydroxymethyl)-p-cresol, tech. 90%
CAS: 91-04-3 | C9H12O3 | 168.192 g/mol
$84.56 - $1147.45
Chemical Identifiers
| CAS | 91-04-3 |
|---|---|
| Molecular Formula | C9H12O3 |
| Molecular Weight (g/mol) | 168.192 |
| MDL Number | MFCD00004619 |
| InChI Key | KUMMBDBTERQYCG-UHFFFAOYSA-N |
| Synonym | 2,6-bis hydroxymethyl-4-methylphenol, 2,6-bis hydroxymethyl-p-cresol, 2,6-dimethylol-p-cresol, 2-hydroxy-5-methyl-1,3-phenylene dimethanol, 2,6-dimethylol-4-methylphenol, 2,6-di hydroxymethyl-p-cresol, 1,3-benzenedimethanol, 2-hydroxy-5-methyl, 2-hydroxy-5-methyl-1,3-benzenedimethanol, 3,5-bis hydroxymethyl-p-cresol, alpha1,alpha,2-trihydroxymesitylene |
| PubChem CID | 7039 |
| IUPAC Name | 2,6-bis(hydroxymethyl)-4-methylphenol |
| SMILES | CC1=CC(=C(C(=C1)CO)O)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1736914
|
Thermo Scientific Chemicals
A1736914 |
25 g |
Each for $84.56
|
|
|||||
|
AAA1736922
|
Thermo Scientific Chemicals
A1736922 |
100 g |
Each for $293.04
|
|
|||||
|
AAA1736936
|
Thermo Scientific Chemicals
A1736936 |
500 g |
Each for $1,147.45
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 91-04-3 | |
| 168.192 | |
| KUMMBDBTERQYCG-UHFFFAOYSA-N | |
| 7039 | |
| CC1=CC(=C(C(=C1)CO)O)CO |
| C9H12O3 | |
| MFCD00004619 | |
| 2,6-bis hydroxymethyl-4-methylphenol, 2,6-bis hydroxymethyl-p-cresol, 2,6-dimethylol-p-cresol, 2-hydroxy-5-methyl-1,3-phenylene dimethanol, 2,6-dimethylol-4-methylphenol, 2,6-di hydroxymethyl-p-cresol, 1,3-benzenedimethanol, 2-hydroxy-5-methyl, 2-hydroxy-5-methyl-1,3-benzenedimethanol, 3,5-bis hydroxymethyl-p-cresol, alpha1,alpha,2-trihydroxymesitylene | |
| 2,6-bis(hydroxymethyl)-4-methylphenol |
Specifications
| 91-04-3 | |
| C9H12O3 | |
| 25 g | |
| 2,6-bis hydroxymethyl-4-methylphenol, 2,6-bis hydroxymethyl-p-cresol, 2,6-dimethylol-p-cresol, 2-hydroxy-5-methyl-1,3-phenylene dimethanol, 2,6-dimethylol-4-methylphenol, 2,6-di hydroxymethyl-p-cresol, 1,3-benzenedimethanol, 2-hydroxy-5-methyl, 2-hydroxy-5-methyl-1,3-benzenedimethanol, 3,5-bis hydroxymethyl-p-cresol, alpha1,alpha,2-trihydroxymesitylene | |
| CC1=CC(=C(C(=C1)CO)O)CO | |
| 168.192 | |
| 168.19 | |
| Technical |
| 119°C to 126°C | |
| MFCD00004619 | |
| 1240237 | |
| KUMMBDBTERQYCG-UHFFFAOYSA-N | |
| 2,6-bis(hydroxymethyl)-4-methylphenol | |
| 7039 | |
| 90% | |
| 2,6-Bis(hydroxymethyl)-p-cresol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
EINECSNumber : 202-036-4
RTECSNumber : CZ6440000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only