accessibility menu, dialog, popup

UserName

CAS RN 91-04-3

IUPAC Name: 2,6-bis(hydroxymethyl)-4-methylphenol
Synonyms: 2,6-bis(hydroxymethyl)-4-methylphenol
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: KUMMBDBTERQYCG-UHFFFAOYSA-N
SMILES: CC1=CC(CO)=C(O)C(CO)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)