missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(hydroxymethyl)-p-cresol, tech. 90%

Catalog No. AAA1736914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1736914 25 g
AAA1736922 100 g
AAA1736936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1736914 Supplier Thermo Scientific Chemicals Supplier No. A1736914

CAS: 91-04-3 | C9H12O3 | 168.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91-04-3
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004619
InChI Key KUMMBDBTERQYCG-UHFFFAOYSA-N
Synonym 2,6-bis hydroxymethyl-4-methylphenol, 2,6-bis hydroxymethyl-p-cresol, 2,6-dimethylol-p-cresol, 2-hydroxy-5-methyl-1,3-phenylene dimethanol, 2,6-dimethylol-4-methylphenol, 2,6-di hydroxymethyl-p-cresol, 1,3-benzenedimethanol, 2-hydroxy-5-methyl, 2-hydroxy-5-methyl-1,3-benzenedimethanol, 3,5-bis hydroxymethyl-p-cresol, alpha1,alpha,2-trihydroxymesitylene
PubChem CID 7039
IUPAC Name 2,6-bis(hydroxymethyl)-4-methylphenol
SMILES CC1=CC(=C(C(=C1)CO)O)CO

Specifications

Melting Point 119°C to 126°C
Quantity 25 g
Beilstein 1240237
Formula Weight 168.19
Percent Purity 90%
Grade Technical
Chemical Name or Material 2,6-Bis(hydroxymethyl)-p-cresol
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-036-4
RTECSNumber CZ6440000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only