missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(hydroxymethyl)-p-cresol 98.0+%, TCI America™

Catalog No. B152525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B152525G 25 g
B1525250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B152525G Supplier TCI America Supplier No. B152525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91-04-3
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004619
InChI Key KUMMBDBTERQYCG-UHFFFAOYSA-N
Synonym 2,6-bis hydroxymethyl-4-methylphenol, 2,6-bis hydroxymethyl-p-cresol, 2,6-dimethylol-p-cresol, 2-hydroxy-5-methyl-1,3-phenylene dimethanol, 2,6-dimethylol-4-methylphenol, 2,6-di hydroxymethyl-p-cresol, 1,3-benzenedimethanol, 2-hydroxy-5-methyl, 2-hydroxy-5-methyl-1,3-benzenedimethanol, 3,5-bis hydroxymethyl-p-cresol, alpha1,alpha,2-trihydroxymesitylene
PubChem CID 7039
IUPAC Name 2,6-bis(hydroxymethyl)-4-methylphenol
SMILES CC1=CC(=C(C(=C1)CO)O)CO

Specifications

Melting Point 126°C
Quantity 25 g
Formula Weight 168.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(hydroxymethyl)-p-cresol
RTECSNumber CZ6440000
TSCA Yes