missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(hydroxymethyl)-1,4-dimethoxybenzene 97.0+%, TCI America™

Catalog No. B26181G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26181G Supplier TCI America Supplier No. B26181G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78840-04-7
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
InChI Key AEZFHUSCRNLFCA-UHFFFAOYSA-N
PubChem CID 13938528
IUPAC Name [3-(hydroxymethyl)-2,5-dimethoxyphenyl]methanol
SMILES COC1=CC(=C(C(=C1)CO)OC)CO

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 1 g
Formula Weight 198.22
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(hydroxymethyl)-1,4-dimethoxybenzene