accessibility menu, dialog, popup

UserName

H3COHOOCH3OH

CAS RN 78840-04-7

IUPAC Name: [3-(hydroxymethyl)-2,5-dimethoxyphenyl]methanol
Synonyms: [3-(hydroxymethyl)-2,5-dimethoxyphenyl]methanol
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: AEZFHUSCRNLFCA-UHFFFAOYSA-N
SMILES: COC1=CC(CO)=C(OC)C(CO)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results