missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Bis(chloromethyl)-1,4-dimethoxybenzene, TCI America™
Supplier: TCI America B26491G
Specifications
| 2,6-Bis(chloromethyl)-1,4-dimethoxybenzene | |
| White-Yellow | |
| 1 g | |
| COC1=CC(=C(C(=C1)CCl)OC)CCl | |
| 235.104 | |
| 235.10 |
| 1379163-23-1 | |
| C10H12Cl2O2 | |
| YMYUSUWLRACOTR-UHFFFAOYSA-N | |
| 1,3-bis(chloromethyl)-2,5-dimethoxybenzene | |
| 44629789 | |
| Crystalline Powder |
Chemical Identifiers
| 1379163-23-1 | |
| 235.104 | |
| 44629789 | |
| COC1=CC(=C(C(=C1)CCl)OC)CCl |
| C10H12Cl2O2 | |
| YMYUSUWLRACOTR-UHFFFAOYSA-N | |
| 1,3-bis(chloromethyl)-2,5-dimethoxybenzene |
Safety and Handling
TSCA : No