accessibility menu, dialog, popup

UserName

CAS RN 1379163-23-1

H3COClOCH3Cl

CAS RN 1379163-23-1

IUPAC Name: 1,3-bis(chloromethyl)-2,5-dimethoxybenzene
Synonyms: 1,3-bis(chloromethyl)-2,5-dimethoxybenzene
Molecular Weight (g/mol): 235.1
Molecular Formula: C10H12Cl2O2
InChi Key: YMYUSUWLRACOTR-UHFFFAOYSA-N
SMILES: COC1=CC(CCl)=C(OC)C(CCl)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results