missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol 98.0+%, TCI America™

Catalog No. B197225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B197225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B197225G Supplier TCI America Supplier No. B197225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1620-68-4
Molecular Formula C23H24O3
Molecular Weight (g/mol) 348.44
MDL Number MFCD00155175
InChI Key MAQOZOILPAMFSW-UHFFFAOYSA-N
Synonym 2,6-Bis(p-cresol-2-ylmethyl)-p-cresol
PubChem CID 74181
IUPAC Name 2,6-bis[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
SMILES CC1=CC=C(O)C(CC2=CC(C)=CC(CC3=CC(C)=CC=C3O)=C2O)=C1

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 25 g
Formula Weight 348.44
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol