CAS RN 1620-68-4
IUPAC Name:
2,6-bis[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
Synonyms:
2,6-bis[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
Molecular Weight (g/mol):
348.44
Molecular Formula:
C23H24O3
InChi Key:
MAQOZOILPAMFSW-UHFFFAOYSA-N
SMILES:
CC1=CC=C(O)C(CC2=CC(C)=CC(CC3=CC(C)=CC=C3O)=C2O)=C1
1
–
1
of
1
results
1
2,6-Bis[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol 98.0+%, TCI America™
CAS: 1620-68-4 Molecular Formula: C23H24O3 Molecular Weight (g/mol): 348.44 MDL Number: MFCD00155175 InChI Key: MAQOZOILPAMFSW-UHFFFAOYSA-N Synonym: 2,6-Bis(p-cresol-2-ylmethyl)-p-cresol PubChem CID: 74181 IUPAC Name: 2,6-bis[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol SMILES: CC1=CC=C(O)C(CC2=CC(C)=CC(CC3=CC(C)=CC=C3O)=C2O)=C1