missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Thiophenedicarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America T234725G
Specifications
| 2,5-Thiophenedicarboxylic Acid | |
| 359°C | |
| C6H4O4S | |
| 25 g | |
| YCGAZNXXGKTASZ-UHFFFAOYSA-N | |
| thiophene-2,5-dicarboxylic acid | |
| 20273 | |
| ≥98.0% (GC,T) |
| 4282-31-9 | |
| White-Yellow | |
| MFCD00016896 | |
| 2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid | |
| C1=C(SC(=C1)C(=O)O)C(=O)O | |
| 172.154 | |
| 172.15 | |
| Crystalline Powder |
Chemical Identifiers
| 4282-31-9 | |
| 172.154 | |
| YCGAZNXXGKTASZ-UHFFFAOYSA-N | |
| 20273 | |
| C1=C(SC(=C1)C(=O)O)C(=O)O |
| C6H4O4S | |
| MFCD00016896 | |
| 2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid | |
| thiophene-2,5-dicarboxylic acid |
Safety and Handling
TSCA : Yes