accessibility menu, dialog, popup

UserName

CAS RN 4282-31-9

IUPAC Name: thiophene-2,5-dicarboxylic acid
Synonyms: 2,5-thiophenedicarboxylic acid
Molecular Weight (g/mol): 172.15
Molecular Formula: C6H4O4S
InChi Key: YCGAZNXXGKTASZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(S1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)